4-Fluoro-2-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

005198-25G 005198-100G
FLUO005198-25GEA 0 GBP
FLUO005198-25G FLUO005198-100G
4-Fluoro-2-nitrotoluene
4-Fluoro-2-nitrotoluene
Formula: C₇H₆FNO₂
Boiling Pt: 214 °C (1013 hPa)
Melting Pt: 27 °C
Density: 1.2911 g/cm³ (0 °C)
MDL Number: MFCD00007200
CAS Number: 446-10-6
EINECS: 207-161-8
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR