3-Fluoro-4-nitrotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
002352-5G
002352-25G
002352-100G
FLUO002352-5GEA
0
GBP
FLUO002352-5G
FLUO002352-25G
FLUO002352-100G
3-Fluoro-4-nitrotoluene
3-Fluoro-4-nitrotoluene
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 249 °C (1013 hPa) Melting Pt: 53 °C Density: 1.438 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00007053 CAS Number: 446-34-4 EINECS: 207-166-5 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...