2-Fluoro-6-nitrotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
009561-1G
009561-100G
009561-25G
FLUO009561-1GEA
0
GBP
FLUO009561-1G
FLUO009561-100G
FLUO009561-25G
2-Fluoro-6-nitrotoluene
2-Fluoro-6-nitrotoluene
Formula:
C₇H₆FNO₂ MW: 155.13 g/mol Boiling Pt: 213 °C (1013 hPa) Melting Pt: 6 °C Density: 1.2766 g/cm³ (20 °C) |
MDL Number:
MFCD00007197 CAS Number: 769-10-8 EINECS: 212-203-3 UN: 2810 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...