α,α,α-Trifluoro-4-iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Iodobenzotrifluoride, 1-Iodo-4-(trifluoromethyl)benzene, α,α,α-Trifluoro-p-iodotoluene

CFF651.5 CFF651.25 001196.25G 001196-100G
CHMPCFF651.5EA 0 GBP
CHMPCFF651.5 CHMPCFF651.25 FLUO001196.25G FLUO001196-100G
α,α,α-Trifluoro-4-iodotoluene
α,α,α-Trifluoro-4-iodotoluene
Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 183…185 °C (1013 hPa)
Melting Pt: 17 °C
Density: 1,851 g/cm³ (26 °C)
MDL Number: MFCD00039398
CAS Number: 455-13-0
EINECS: 207-234-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR