α,α,α-Trifluoro-3-iodotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: α,α,α-Trifluoro-m-iodotoluene, 3-Iodobenzotrifluoride, 1-Iodo-3-(trifluoromethyl)benzene

001195.25G 001195.100G
FLUO001195.25GEA 0 GBP
FLUO001195.25G FLUO001195.100G
α,α,α-Trifluoro-3-iodotoluene
α,α,α-Trifluoro-3-iodotoluene
Formula: C₇H₄F₃I
MW: 272.01 g/mol
Boiling Pt: 182 °C (1013 hPa)
Melting Pt: –8 °C
Density: 1.887 g/cm³ (20 °C)
MDL Number: MFCD00001049
CAS Number: 401-81-0
EINECS: 206-934-7
UN: 3265
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR