(4-Fluorophenyl)acetone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

009228-10G 009228-1G 009228-5G
FLUO009228-10GEA 0 GBP
FLUO009228-10G FLUO009228-1G FLUO009228-5G
(4-Fluorophenyl)acetone
(4-Fluorophenyl)acetone
Formula: C₉H₉FO
Boiling Pt: 106 °C (1013 hPa)
Density: 1.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000362
CAS Number: 459-03-0
EINECS: 207-284-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR