4-Fluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Fluoro-4-methylbenzene
001443-250G
001443-1KG
FLUO001443-250GEA
0
GBP
FLUO001443-250G
FLUO001443-1KG
4-Fluorotoluene
4-Fluorotoluene
Formula:
C₇H₇F MW: 110.13 g/mol Boiling Pt: 116 Melting Pt: –57 °C Density: 1 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000358 CAS Number: 352-32-9 EINECS: 206-520-6 UN: 2388 ADR: 3 II Merck Index: 13,04207 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...