2-Fluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-Fluoro-2-methylbenzene, o-Fluorotoluene
001423-250G
001423-1KG
FLUO001423-250GEA
0
GBP
FLUO001423-250G
FLUO001423-1KG
2-Fluorotoluene
2-Fluorotoluene
Formula:
C₇H₇F MW: 110.13 g/mol Boiling Pt: 113 Melting Pt: –62 °C Density: 1.005 g/cm³ (20 °C) Flash Pt: 8 Storage Temperature: Ambient |
MDL Number:
MFCD00000322 CAS Number: 95-52-3 EINECS: 202-428-5 UN: 2388 ADR: 3 II Merck Index: 13,04207 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...