2,5-Difluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,4-Difluoro-2-methylbenzene

001466-25G 001466-10G
FLUO001466-25GEA 0 GBP
FLUO001466-25G FLUO001466-10G
2,5-Difluorotoluene
2,5-Difluorotoluene
Formula: C₇H₆F₂
MW: 128.12 g/mol
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.36 g/cm³ (20 °C)
MDL Number: MFCD00000347
CAS Number: 452-67-5
EINECS: 207-205-6
UN: 2388
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR