5-Chloro-2-nitrobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Chloro-1-nitro-2-(trifluoromethyl)benzene, 5-Chloro-2-nitrobenzotrifluoride, 1-Chloro-4-nitro-3-(trifluoromethyl)benzene, 5-Chloro-α,α,α-trifluoro-2-nitrotoluene

004509-25G 004509-100G
FLUO004509-25GEA 0 GBP
FLUO004509-25G FLUO004509-100G
5-Chloro-2-nitrobenzotrifluoride
5-Chloro-2-nitrobenzotrifluoride
Formula: C₇H₃ClF₃NO₂
MW: 225.55 g/mol
Boiling Pt: 222 °C (1013 hPa)
Melting Pt: 21…22 °C
Density: 1.526 g/cm³ (20 °C)
MDL Number: MFCD00007298
CAS Number: 118-83-2
EINECS: 204-280-7
UN: 2810
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR