3-Fluoro-α,α,α-trichlorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3-Fluorobenzotrichloride, m-Fluoro-α,α,α-trichlorotoluene
010681-1G
010681-5G
FLUO010681-1GEA
0
GBP
FLUO010681-1G
FLUO010681-5G
3-Fluoro-α,α,α-trichlorotoluene
3-Fluoro-α,α,α-trichlorotoluene
Formula:
C₇H₄Cl₃F MW: 213.47 g/mol Boiling Pt: 119.4 °C (45 mmHg) Density: 1,4408 g/cm³ (30 °C) |
MDL Number:
MFCD00042073 CAS Number: 401-77-4 EINECS: 206-931-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...