2-Chloro-5-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Chloro-4-fluoro-2-methylbenzene

001598-5G 001598-25G
FLUO001598-5GEA 0 GBP
FLUO001598-5G FLUO001598-25G
2-Chloro-5-fluorotoluene
2-Chloro-5-fluorotoluene
Formula: C₇H₆ClF
Boiling Pt: 156 °C (1013 hPa)
Density: 1,18 g/cm³ (20 °C)
MDL Number: MFCD00040853
CAS Number: 33406-96-1
EINECS: 251-508-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR