2-Chloro-4-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-4-fluoro-1-methylbenzene

001597-1KG 001597-250G
FLUO001597-1KGEA 0 GBP
FLUO001597-1KG FLUO001597-250G
2-Chloro-4-fluorotoluene
2-Chloro-4-fluorotoluene
Formula: C₇H₆ClF
Boiling Pt: 151.5…153 °C (1013 hPa)
Density: 1.1972 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000572
CAS Number: 452-73-3
EINECS: 207-209-8
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR