4-Chloro-2-fluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001672-100G
001672-250G
FLUO001672-100GEA
0
GBP
FLUO001672-100G
FLUO001672-250G
4-Chloro-2-fluorotoluene
4-Chloro-2-fluorotoluene
Formula:
C₇H₆ClF Boiling Pt: 153 °C (1013 hPa) Density: 1,184 g/cm³ (20 °C) |
MDL Number:
MFCD00000571 CAS Number: 452-75-5 EINECS: 207-210-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...