2-Chloro-6-fluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
001599-100G
001599.250G
001599-500G
FLUO001599-100GEA
0
GBP
FLUO001599-100G
FLUO001599.250G
FLUO001599-500G
2-Chloro-6-fluorotoluene
2-Chloro-6-fluorotoluene
Formula:
C₇H₆ClF Boiling Pt: 154 °C (1013 hPa) Melting Pt: –27 °C Density: 1.19 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000570 CAS Number: 443-83-4 EINECS: 207-141-9 UN: 1993 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...