4-Fluorobenzyl chloride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
4-Fluorobenzyl chloride, p-Fluorobenzyl chloride, 1-Chloromethyl-4-fluorobenzene, α-Chloro-4-fluorotoluene, 1-(Chloromethyl)-4-fluorobenzene
006201-100G
006201-500G
FLUO006201-100GEA
0
GBP
FLUO006201-100G
FLUO006201-500G
4-Fluorobenzyl chloride
4-Fluorobenzyl chloride
Formula:
C₇H₆ClF MW: 144.58 g/mol Boiling Pt: 181.2 Melting Pt: –18 °C Density: 1.21 g/cm³ (20 °C) Flash Pt: 60 Storage Temperature: Ambient |
MDL Number:
MFCD00000913 CAS Number: 352-11-4 EINECS: 206-516-4 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...