α-Chloro-3-fluorotoulene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-(Chloromethyl)-3-fluorobenzene, α-Chloro-m-fluorotoulene, 3-Fluorobenzyl chloride
001632-25G
001632-100G
FLUO001632-25GEA
0
GBP
FLUO001632-25G
FLUO001632-100G
α-Chloro-3-fluorotoulene
α-Chloro-3-fluorotoulene
Formula:
C₇H₆ClF Boiling Pt: 67 °C (1013 hPa) Density: 1.21 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000904 CAS Number: 456-42-8 EINECS: 207-264-8 UN: 2920 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...