α-Chloro-2-fluorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 2-Fluorobenzyl chloride, 1-(Chloromethyl)-2-fluorobenzene, α-Chloro-o-fluorotoulene

001422-100G 001422-250G FL1422.1
FLUO001422-100GEA 0 GBP
FLUO001422-100G FLUO001422-250G CHMPFL1422.1
α-Chloro-2-fluorotoulene
α-Chloro-2-fluorotoulene
Formula: C₇H₆ClF
MW: 144.58 g/mol
Boiling Pt: 86
Density: 1.21 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000892
CAS Number: 345-35-7
EINECS: 206-460-0
UN: 2920
ADR: 8 II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR