2,4-Dichlorobenzotrifluoride
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
2,4-Dichlorobenzotrifluoride, 1,3-Dichloro-4-(trifluoromethyl)benzene, 2,4-Dichloro-1-(trifluoromethyl)benzene
004693-100G
004693-500G
004693-1KG
FLUO004693-100GEA
0
GBP
FLUO004693-100G
FLUO004693-500G
FLUO004693-1KG
2,4-Dichlorobenzotrifluoride
2,4-Dichlorobenzotrifluoride
Formula:
C₇H₃Cl₂F₃ MW: 215 g/mol Boiling Pt: 193 °C (1013 hPa) Melting Pt: –1 °C Density: 1.4929 g/cm³ (20 °C) |
MDL Number:
MFCD00000582 CAS Number: 320-60-5 EINECS: 206-278-1 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...