3,4-Dichloro-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,2-Dichloro-4-(trifluoromethyl)benzene, 3,4-Dichlorobenzotrifluoride

004694-100G 004694-500G
FLUO004694-100GEA 0 GBP
FLUO004694-100G FLUO004694-500G
3,4-Dichloro-α,α,α-trifluorotoluene
3,4-Dichloro-α,α,α-trifluorotoluene
Formula: C₇H₃Cl₂F₃
MW: 215 g/mol
Boiling Pt: 173
Melting Pt: –0.2 °C
Density: 1.486 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000555
CAS Number: 328-84-7
EINECS: 206-337-1
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR