α,3-Dichlorotoluene ≥97%
Supplier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
1-Chloro-3-(chloromethyl)benzene, 3-Chlorobenzyl chloride, 3-Chlorobenzylchloride
A11041.14
A11041.22
AB114692.0025
A11041.14EA
26.6
GBP
A11041.14
A11041.22
ABCRAB114692.0025
α,3-Dichlorotoluene ≥97%
α,3-Dichlorotoluene
Purity: 97%
CAS No.: 620-20-2
Molecular Formula: C7H6Cl2
Formula Weight: 161.03
Flash Point: 98°(208°F)
Refractive Index: 1.5560
Odour: Pungent
Boiling Point: 215-216°
Beilstein Reference: 742266
UN No.: 2235
MDL No.: MFCD00000905
Formula:
C₇H₆Cl₂ MW: 161.03 g/mol Boiling Pt: 215…216 °C Melting Pt: 69…71 °C Density: 1.270 Flash Pt: 98 °C (208 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000905 CAS Number: 620-20-2 EINECS: 210-629-4 UN: 2235 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...