2,5-Dichlorotoluene ≥98%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1,4-Dichloro-2-methylbenzene

A13039.14 A13039.22
A13039.14EA 42.6 GBP
A13039.14 A13039.22
2,5-Dichlorotoluene ≥98%
2,5-Dichlorotoluene

Purity: 98% CAS No.: 19398-61-9 Molecular Formula: C7H6Cl2 Formula Weight: 161.03 Flash Point: 79°(174°F) Refractive Index: 1.5470 Melting Point: 4-5° Boiling Point: 198-200° Beilstein Reference: 1859112 UN No.: 3082 MDL No.: MFCD00000609

Formula: C₇H₆Cl₂
MW: 161.03 g/mol
Boiling Pt: 198…200 °C
Melting Pt: 4…5 °C
Density: 1.256
Flash Pt: 79 °C (174 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000609
CAS Number: 19398-61-9
EINECS: 243-032-2
UN: 3082
ADR: 9 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR