4-Amino-m-toluic acid

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Amino-3-methylbenzoic acid

019461-5G 019461-25G
FLUO019461-5GEA 0 GBP
FLUO019461-5G FLUO019461-25G
4-Amino-m-toluic acid
4-Amino-m-toluic acid
Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 340
Melting Pt: 170
MDL Number: MFCD00007736
CAS Number: 2486-70-6
EINECS: 219-629-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR