4-Butylbenzoic acid ≥99%

Supplier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

B23891.06 B23891.14 B23891.22
B23891.06EA 24.1 GBP
B23891.06 B23891.14 B23891.22
4-Butylbenzoic acid ≥99%
4-Butylbenzoic acid

Purity: 99% CAS No.: 20651-71-2 Molecular Formula: C11H14O2 Formula Weight: 178.23 Melting Point: 100-113° MDL No.: MFCD00002571

Formula: C₁₁H₁₄O₂
Boiling Pt: 313 °C (1013 hPa)
Melting Pt: 100…113 °C
Density: ∼1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002571
CAS Number: 20651-71-2
EINECS: 243-940-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR