3-Methoxyphenylacetone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
1-(3-Methoxyphenyl)propan-2-one
018796-1G
018796-5G
FLUO018796-1GEA
0
GBP
FLUO018796-1G
FLUO018796-5G
3-Methoxyphenylacetone
3-Methoxyphenylacetone
Formula:
C₁₀H₁₂O₂ |
MDL Number:
MFCD00008771 CAS Number: 3027-13-2 EINECS: 221-191-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...