3-Tolualdehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: m-Tolualdehyde, 3-Methylbenzaldehyde, m-Tolylaldehyde

001390-25G 001390-100G
FLUO001390-25GEA 0 GBP
FLUO001390-25G FLUO001390-100G
3-Tolualdehyde
3-Tolualdehyde
Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 199 °C (1013 hPa)
Density: 1.0535 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003374
CAS Number: 620-23-5
EINECS: 210-632-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR