2-Tolualdehyde

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

001389-25G 001389-100G 001389-500G
FLUO001389-25GEA 0 GBP
FLUO001389-25G FLUO001389-100G FLUO001389-500G
2-Tolualdehyde
2-Tolualdehyde
Formula: C₈H₈O
Boiling Pt: 201 °C (1013 hPa)
Melting Pt: –35 °C
Density: 1.0324 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003338
CAS Number: 529-20-4
EINECS: 208-452-2
Merck Index: 12,09665

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR