2-Bromo-4-methylanisole

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

018292-10G 018292-25G 018292-5G
FLUO018292-10GEA 0 GBP
FLUO018292-10G FLUO018292-25G FLUO018292-5G
2-Bromo-4-methylanisole
2-Bromo-4-methylanisole
Formula: C₈H₉BrO
MDL Number: MFCD00017783
CAS Number: 22002-45-5
EINECS: 244-705-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR