4-Bromo-3-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001728-25G 001728-100G
FLUO001728-25GEA 0 GBP
FLUO001728-25G FLUO001728-100G
4-Bromo-3-nitrotoluene
4-Bromo-3-nitrotoluene
Formula: C₇H₆BrNO₂
Boiling Pt: 84…100 °C (0.25 mmHg)
Melting Pt: 35 °C
Density: 1,57 g/cm³ (20 °C)
MDL Number: MFCD00024180
CAS Number: 5326-34-1
EINECS: 226-203-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR