3-Bromo-2-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

010655-25G 010655-5G
FLUO010655-25GEA 0 GBP
FLUO010655-25G FLUO010655-5G
3-Bromo-2-nitrotoluene
3-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
Boiling Pt: 237 °C (1013 hPa)
Melting Pt: 25…29 °C
Density: 1.612 g/cm³ (25 °C)
MDL Number: MFCD02673631
CAS Number: 52414-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR