2-Bromo-4-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001370-25G 001370-100G
FLUO001370-25GEA 0 GBP
FLUO001370-25G FLUO001370-100G
2-Bromo-4-nitrotoluene
2-Bromo-4-nitrotoluene
Formula: C₇H₆BrNO₂
Boiling Pt: 175 °C (38 mmHg)
Melting Pt: 77,5 °C
MDL Number: MFCD00007195
CAS Number: 7745-93-9
EINECS: 231-809-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR