α-Bromo-2-nitrotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002679-5G 002679-25G
FLUO002679-5GEA 0 GBP
FLUO002679-5G FLUO002679-25G
α-Bromo-2-nitrotoluene
α-Bromo-2-nitrotoluene
Formula: C₇H₆BrNO₂
Melting Pt: 45…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007184
CAS Number: 3958-60-9
EINECS: 223-558-9
UN: 3261
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR