4-Bromo-3-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 5-Amino-2-bromobenzotrifluoride, 4-Bromo-α,α,α-trifluoro-m-toluidine, 5-Amino-2-bromo-α,α,α-trifluorotoluene

CFF424.10 CFF424.25 CFF424.50 002244-50G 002244-250G
CHMPCFF424.10EA 0 GBP
CHMPCFF424.10 CHMPCFF424.25 CHMPCFF424.50 FLUO002244-50G FLUO002244-250G
4-Bromo-3-(trifluoromethyl)aniline
4-Bromo-3-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 81…84 °C (0.5 mmHg)
Melting Pt: 55…56 °C
MDL Number: MFCD00007827
CAS Number: 393-36-2
EINECS: 206-885-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR