4-Bromo-2-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-α,α,α-trifluoro-o-toluidine, 2-Amino-5-bromobenzotrifluoride, 2-Amino-5-bromo-α,α,α-trifluorotoluene, 2-Amino-5-bromo-α,α,α-trifluortoluene

001983-25G 001983-100G
FLUO001983-25GEA 0 GBP
FLUO001983-25G FLUO001983-100G
4-Bromo-2-(trifluoromethyl)aniline
4-Bromo-2-(trifluoromethyl)aniline
Formula: C₇H₅BrF₃N
MW: 240.02 g/mol
Boiling Pt: 60.7…62 °C (1 mmHg)
Density: 1.712 g/cm³ (25 °C)
MDL Number: MFCD00064393
CAS Number: 445-02-3
EINECS: 207-150-8
UN: 2810
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR