2,6-Dibromo-4-(trifluoromethyl)aniline

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

CFF546.10 004680-1G 004680-5G
CHMPCFF546.10EA 0 GBP
CHMPCFF546.10 FLUO004680-1G FLUO004680-5G
2,6-Dibromo-4-(trifluoromethyl)aniline
2,6-Dibromo-4-(trifluoromethyl)aniline
Formula: C₇H₄Br₂F₃N
Boiling Pt: 64…65 °C (0.1 mmHg)
Melting Pt: 38…40 °C
MDL Number: MFCD00068181
CAS Number: 72678-19-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR