5-Bromo-2-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 4-Bromo-1-fluoro-2-methylbenzene

CFF579.10 001620-25G 001620-100G 001620-500G
CHMPCFF579.10EA 0 GBP
CHMPCFF579.10 FLUO001620-25G FLUO001620-100G FLUO001620-500G
5-Bromo-2-fluorotoluene
5-Bromo-2-fluorotoluene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 94…95 °C (66.7 hPa)
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000343
CAS Number: 51437-00-4
EINECS: 257-202-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR