2-Bromo-5-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

001613-25G 001613-100G
FLUO001613-25GEA 0 GBP
FLUO001613-25G FLUO001613-100G
2-Bromo-5-fluorotoluene
2-Bromo-5-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 177 °C (1013 hPa)
Density: 1.49 g/cm³ (20 °C)
MDL Number: MFCD00017921
CAS Number: 452-63-1
EINECS: 207-203-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR