4-Bromo-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

Synonyms: 4-Bromobenzotrifluoride, 1-Bromo-4-(trifluoromethyl)benzene

002195-100G 002195-500G 002195-1KG
FLUO002195-100GEA 0 GBP
FLUO002195-100G FLUO002195-500G FLUO002195-1KG
4-Bromo-α,α,α-trifluorotoluene
4-Bromo-α,α,α-trifluorotoluene
Formula: C₇H₄BrF₃
Boiling Pt: 154 °C (1013 hPa)
Density: 1.6347 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000398
CAS Number: 402-43-7
EINECS: 206-943-6
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR