2-Bromo-4-fluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

001605-25G 001605-100G
FLUO001605-25GEA 0 GBP
FLUO001605-25G FLUO001605-100G
2-Bromo-4-fluorotoluene
2-Bromo-4-fluorotoluene
Formula: C₇H₆BrF
Boiling Pt: 170…172 °C (1013 hPa)
Density: 1,52 g/cm³ (20 °C)
MDL Number: MFCD00040828
CAS Number: 1422-53-3
EINECS: 215-830-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR