3-Bromo-4-fluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
1-Bromo-2-fluoro-5-methylbenzene
004234-10G
004234-100G
004234-25G
FLUO004234-10GEA
0
GBP
FLUO004234-10G
FLUO004234-100G
FLUO004234-25G
3-Bromo-4-fluorotoluene
3-Bromo-4-fluorotoluene
Formula:
C₇H₆BrF MW: 189.03 g/mol Boiling Pt: 186 Density: 1.5 g/cm³ (20 °C) |
MDL Number:
MFCD00040827 CAS Number: 452-62-0 EINECS: 207-201-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...