α-Bromo-4-fluorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002963-25G 002963-100G 002963-500G
FLUO002963-25GEA 0 GBP
FLUO002963-25G FLUO002963-100G FLUO002963-500G
α-Bromo-4-fluorotoulene
α-Bromo-4-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 85 °C (1013 hPa)
Density: 1.517 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000359
CAS Number: 459-46-1
EINECS: 207-291-5
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR