α-Bromo-3-fluorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Danger

002171-100G 002171-25G
FLUO002171-100GEA 0 GBP
FLUO002171-100G FLUO002171-25G
α-Bromo-3-fluorotoulene
α-Bromo-3-fluorotoulene
Formula: C₇H₆BrF
Boiling Pt: 88 °C (1013 hPa)
Density: 1.541 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00000340
CAS Number: 456-41-7
EINECS: 207-263-2
UN: 3265
ADR: 8,II

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR