α-Bromo-3,4-difluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
3,4-Difluorobenzyl bromide, 4-(Bromomethyl)-1,2-difluorobenzene
002085-10G
002085-25G
002085-100G
FLUO002085-10GEA
0
GBP
FLUO002085-10G
FLUO002085-25G
FLUO002085-100G
α-Bromo-3,4-difluorotoluene
α-Bromo-3,4-difluorotoluene
Formula:
C₇H₅BrF₂ MW: 207.02 g/mol Boiling Pt: 59…61 °C (3.5 mmHg) Density: 1.61 g/cm³ (20 °C) |
MDL Number:
MFCD00009889 CAS Number: 85118-01-0 EINECS: 285-653-1 UN: 3265 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...