α-Bromo-2-fluorotoulene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluorobenzyl bromide, α-Bromo-o-fluorotoulene

006200-25G 006200-100G 006200-250G
FLUO006200-25GEA 0 GBP
FLUO006200-25G FLUO006200-100G FLUO006200-250G
α-Bromo-2-fluorotoulene
α-Bromo-2-fluorotoulene
Formula: C₇H₆BrF
MW: 189.03 g/mol
Boiling Pt: 84
Density: 1.567 g/cm³ (20 °C)
MDL Number: MFCD00000324
CAS Number: 446-48-0
EINECS: 207-169-1
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR