α-Bromo-2,6-difluorotoluene
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Danger
|
Synonyms:
2-(Bromomethyl)-1,3-difluorobenzene, 2,6-Difluorobenzyl bromide
001962-25G
001962-10G
FLUO001962-25GEA
0
GBP
FLUO001962-25G
FLUO001962-10G
α-Bromo-2,6-difluorotoluene
α-Bromo-2,6-difluorotoluene
Formula:
C₇H₅BrF₂ Melting Pt: 53 °C Density: 1.6 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00000329 CAS Number: 85118-00-9 EINECS: 285-652-6 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...