α-Bromo-2,5-difluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(Bromomethyl)-1,4-difluorobenzene, 2,5-Difluorobenzyl bromide

FL4797.1 004797-25G 004797-10G 004797-100G
CHMPFL4797.1EA 0 GBP
CHMPFL4797.1 FLUO004797-25G FLUO004797-10G FLUO004797-100G
α-Bromo-2,5-difluorotoluene
α-Bromo-2,5-difluorotoluene
Formula: C₇H₅BrF₂
Boiling Pt: 186 °C (1013 hPa)
Melting Pt: 8 °C
Density: 1.609 g/cm³ (20 °C)
MDL Number: MFCD00009897
CAS Number: 85117-99-3
EINECS: 285-651-0
UN: 3265
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR