2,5-Dibromo-α,α,α-trifluorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1,4-Dibromo-2-(trifluoromethyl)benzene, 2,5-Dibromobenzotrifluoride

009124-5G 009124-25G
FLUO009124-5GEA 0 GBP
FLUO009124-5G FLUO009124-25G
2,5-Dibromo-α,α,α-trifluorotoluene
2,5-Dibromo-α,α,α-trifluorotoluene
Formula: C₇H₃Br₂F₃
MDL Number: MFCD00013554
CAS Number: 7657-09-2
EINECS: 231-620-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR