3-Bromo-4-chlorobenzotrifluoride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-2-chloro-5-(trifluoromethyl)benzene, 2-Bromo-1-chloro-4-(trifluoromethyl)benzene, 3-Bromo-4-chloro-α,α,αa-trifluorotoluene

010672-25G 010672-100G
FLUO010672-25GEA 0 GBP
FLUO010672-25G FLUO010672-100G
3-Bromo-4-chlorobenzotrifluoride
3-Bromo-4-chlorobenzotrifluoride
Formula: C₇H₃BrClF₃
MW: 259.45 g/mol
Boiling Pt: 188 °C (1013 hPa)
Melting Pt: –0.6 °C
Density: 1,743 g/cm³ (25 °C)
MDL Number: MFCD00018093
CAS Number: 454-78-4
EINECS: 207-226-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR