4-Bromobenzyl chloride

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Bromo-α-chlorotoluene, α-Chloro-4-bromotoluene, 1-Bromo-4-(chloromethyl)benzene, 1-(Chloromethyl)-4-bromobenzene

002483-5G 002483-25G
FLUO002483-5GEA 0 GBP
FLUO002483-5G FLUO002483-25G
4-Bromobenzyl chloride
4-Bromobenzyl chloride
Formula: C₇H₆BrCl
MW: 205.48 g/mol
Boiling Pt: 236 °C (1013 hPa)
Melting Pt: 40…41 °C
MDL Number: MFCD00040867
CAS Number: 589-17-3
EINECS: 209-638-6
UN: 3261
ADR: 8,II
Merck Index: 13,01394

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR