2-Bromo-6-chlorotoluene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 1-Bromo-3-chloro-2-methylbenzene

018260-10G 018260-50G
FLUO018260-10GEA 0 GBP
FLUO018260-10G FLUO018260-50G
2-Bromo-6-chlorotoluene
2-Bromo-6-chlorotoluene
Formula: C₇H₆BrCl
MDL Number: MFCD00061121
CAS Number: 62356-27-8
EINECS: 263-520-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR